Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gclm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016536.2 | NP_001016536.1 | 5 - 6 | 667 | 95 | |
GenBank mRNA | BC167312.1 | - | 5 - 6 | 676 | 95 | |
CR760783.2 | - | 5 - 6 | 667 | 95 | ||
CR926228.1 | - | 5 - 6 | 686 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |