Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016594.2 | NP_001016594.1 | 2 - 3 | 326 | 76 | |
GenBank mRNA | BC135275.1 | - | 2 - 3 | 319 | 76 | |
CR760592.2 | - | 2 - 3 | 326 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |