Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mat2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016828.3 | NP_001016828.2 | 5 - 6 | 773 | 90 | |
GenBank mRNA | BC093462.1 | - | 5 - 6 | 773 | 90 | |
CR855516.2 | - | 5 - 6 | 792 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |