Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crnkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016895.2 | NP_001016895.1 | 3 - 4 | 328 | 71 | |
GenBank mRNA | BC155382.1 | - | 3 - 4 | 307 | 71 | |
CR848544.2 | - | 3 - 4 | 328 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |