Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spryd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016934.2 | NP_001016934.1 | 3 - 4 | 389 | 105 | |
XM_012957010.2 | XP_012812464.1 | 4 - 5 | 1045 | 105 | ||
XM_012957011.2 | XP_012812465.1 | 3 - 4 | 395 | 105 | ||
GenBank mRNA | CR762113.2 | - | 3 - 4 | 389 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Xenopus tropicalis Probe spans exons |