Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cmss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016943.2 | NP_001016943.1 | 1 - 2 | 89 | 84 | |
GenBank mRNA | BC135225.1 | - | 1 - 2 | 143 | 84 | |
CR761873.2 | - | 1 - 2 | 89 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |