Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016947.3 | NP_001016947.2 | 3 - 4 | 797 | 78 | |
GenBank mRNA | BC121545.1 | - | 3 - 4 | 768 | 78 | |
BX755136.1 | - | 1 - 2 | 332 | 78 | ||
CR761760.2 | - | 3 - 4 | 795 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |