Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017017.2 | NP_001017017.1 | 2 - 3 | 644 | 93 | |
GenBank mRNA | CR762174.2 | - | 2 - 3 | 644 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |