Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017023.2 | NP_001017023.1 | 4 - 5 | 435 | 83 | |
XM_018092101.1 | XP_017947590.1 | 4 - 5 | 579 | 83 | ||
XM_018092102.1 | XP_017947591.1 | 3 - 4 | 387 | 83 | ||
GenBank mRNA | BC167324.1 | - | 4 - 5 | 533 | 83 | |
CR762087.2 | - | 4 - 5 | 435 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |