Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lztfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017032.2 | NP_001017032.1 | 1 - 2 | 75 | 112 | |
GenBank mRNA | BC135727.1 | - | 1 - 2 | 45 | 112 | |
BC157282.1 | - | 1 - 2 | 95 | 112 | ||
CR762013.2 | - | 1 - 2 | 75 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |