Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lztfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017032.2 | NP_001017032.1 | 4 - 5 | 455 | 113 | |
GenBank mRNA | BC135727.1 | - | 4 - 5 | 421 | 113 | |
BC157282.1 | - | 4 - 5 | 475 | 113 | ||
CR762013.2 | - | 4 - 5 | 455 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |