Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017071.2 | NP_001017071.1 | 3 - 4 | 249 | 78 | |
GenBank mRNA | CR761547.2 | - | 3 - 4 | 249 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Xenopus tropicalis Probe spans exons |