Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfand3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017147.2 | NP_001017147.1 | 159 | 101 | ||
GenBank mRNA | BC157766.1 | - | 203 | 101 | ||
CR760946.2 | - | 159 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis |