Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017191.2 | NP_001017191.1 | 7 - 8 | 889 | 97 | |
GenBank mRNA | BC121552.1 | - | 7 - 8 | 808 | 97 | |
CR760599.2 | - | 7 - 8 | 889 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |