Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qpct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017245.2 | NP_001017245.1 | 5 - 6 | 825 | 123 | |
GenBank mRNA | BC118705.1 | - | 5 - 6 | 869 | 123 | |
CR760147.2 | - | 5 - 6 | 825 | 123 |
Gene Symbol: LOC108644434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018094262.1 | XP_017949751.1 | 5 - 6 | 819 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |