Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pard6g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017338.2 | NP_001017338.1 | 1 - 2 | 174 | 70 | |
GenBank mRNA | CR855473.2 | - | 1 - 2 | 174 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Xenopus tropicalis Probe spans exons |