Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018091310.1 | XP_017946799.1 | 4 - 5 | 1441 | 102 | |
GenBank mRNA | BC135209.1 | - | 3 - 4 | 970 | 102 | |
DQ124206.1 | - | 3 - 4 | 1141 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |