Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctsc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037259.1 | NP_001032336.1 | 3 - 4 | 489 | 104 | |
XM_012956689.2 | XP_012812143.1 | 3 - 4 | 567 | 104 | ||
GenBank mRNA | BC067941.1 | - | 3 - 4 | 515 | 104 | |
BC098085.1 | - | 3 - 4 | 484 | 104 | ||
CR761110.2 | - | 3 - 4 | 489 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |