Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctsc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037259.1 | NP_001032336.1 | 6 - 7 | 892 | 64 | |
XM_012956689.2 | XP_012812143.1 | 6 - 7 | 970 | 64 | ||
GenBank mRNA | BC067941.1 | - | 6 - 7 | 918 | 64 | |
BC098085.1 | - | 6 - 7 | 887 | 64 | ||
CR761110.2 | - | 6 - 7 | 892 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |