Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arpp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037260.1 | NP_001032337.1 | 2 - 3 | 189 | 72 | |
GenBank mRNA | BC075509.1 | - | 2 - 3 | 142 | 72 | |
BC096631.1 | - | 2 - 3 | 195 | 72 | ||
BC123052.1 | - | 2 - 3 | 164 | 72 | ||
CR761222.2 | - | 2 - 3 | 189 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |