Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: akr7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039732.1 | NP_001034821.1 | 485 | 85 | ||
GenBank mRNA | BC076912.1 | - | 476 | 85 | ||
BC171130.1 | - | 2 - 3 | 477 | 85 | ||
CR759999.2 | - | 485 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |