Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039740.1 | NP_001034829.1 | 2 - 3 | 182 | 91 | |
XM_012961021.1 | XP_012816475.1 | 3 - 4 | 308 | 91 | ||
XM_012961023.1 | XP_012816477.1 | 3 - 4 | 310 | 91 | ||
XM_018093032.1 | XP_017948521.1 | 3 - 4 | 308 | 91 | ||
GenBank mRNA | BC089700.1 | - | 2 - 3 | 159 | 91 | |
BC167302.1 | - | 2 - 3 | 164 | 91 | ||
BC170700.1 | - | 2 - 3 | 159 | 91 | ||
BC170702.1 | - | 2 - 3 | 159 | 91 | ||
CR762362.2 | - | 2 - 3 | 182 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |