Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b3gat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030353.2 | NP_001025524.1 | 3 - 4 | 1258 | 85 | |
GenBank mRNA | AJ890843.1 | - | 3 - 4 | 905 | 85 | |
BC121201.1 | - | 3 - 4 | 1258 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |