Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cenpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044442.3 | NP_001037907.2 | 5 - 6 | 721 | 64 | |
NM_001281828.1 | NP_001268757.1 | 5 - 6 | 645 | 64 | ||
XM_012964045.2 | XP_012819499.1 | 5 - 6 | 754 | 64 | ||
GenBank mRNA | BC095911.1 | - | 5 - 6 | 656 | 64 | |
BC131829.1 | - | 5 - 6 | 690 | 64 | ||
BC161023.1 | - | 5 - 6 | 645 | 64 | ||
CR760735.2 | - | 5 - 6 | 644 | 64 | ||
CX497350.2 | - | 5 - 6 | 721 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |