Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c3orf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044448.1 | NP_001037913.1 | 1 - 2 | 148 | 91 | |
GenBank mRNA | BC160453.1 | - | 2 - 3 | 133 | 91 | |
CR760904.2 | - | 1 - 2 | 148 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |