Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hspb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044511.1 | NP_001037976.1 | 1 - 2 | 202 | 68 | |
GenBank mRNA | BC135182.1 | - | 1 - 2 | 188 | 68 | |
CR761848.2 | - | 1 - 2 | 202 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |