Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: map1lc3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044522.1 | NP_001037987.1 | 1 - 2 | 73 | 92 | |
GenBank mRNA | BC155037.1 | - | 1 - 2 | 116 | 92 | |
CR762033.2 | - | 1 - 2 | 73 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |