Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045588.1 | NP_001039053.1 | 2 - 3 | 219 | 76 | |
GenBank mRNA | CR848515.2 | - | 2 - 3 | 219 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |