Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnase2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045608.1 | NP_001039073.1 | 7 - 8 | 949 | 99 | |
XM_012953172.2 | XP_012808626.1 | 7 - 8 | 1032 | 99 | ||
XM_012953173.1 | XP_012808627.1 | 7 - 8 | 1012 | 99 | ||
XM_012953174.2 | XP_012808628.1 | 7 - 8 | 1002 | 99 | ||
GenBank mRNA | BC135708.1 | - | 6 - 7 | 826 | 99 | |
CR855609.1 | - | 7 - 8 | 949 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |