Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nipsnap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045627.1 | NP_001039092.1 | 4 - 5 | 392 | 85 | |
GenBank mRNA | BC154054.1 | - | 4 - 5 | 358 | 85 | |
CR942347.2 | - | 4 - 5 | 392 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |