Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045689.2 | NP_001039154.1 | 4 - 5 | 348 | 101 | |
GenBank mRNA | CR942586.2 | - | 3 - 4 | 277 | 101 | |
CX433740.2 | - | 4 - 5 | 346 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Xenopus tropicalis Probe spans exons |