Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lonrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045693.1 | NP_001039158.1 | 6 - 7 | 1732 | 95 | |
XM_012957170.2 | XP_012812624.1 | 6 - 7 | 1799 | 95 | ||
GenBank mRNA | BC121307.1 | - | 6 - 7 | 1732 | 95 | |
CR942594.2 | - | 6 - 7 | 1647 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |