Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lonrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045693.1 | NP_001039158.1 | 8 - 9 | 1968 | 105 | |
XM_012957170.2 | XP_012812624.1 | 8 - 9 | 2035 | 105 | ||
GenBank mRNA | BC121307.1 | - | 8 - 9 | 1968 | 105 | |
CR942594.2 | - | 8 - 9 | 1883 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |