Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem30b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045712.1 | NP_001039177.1 | 5 - 6 | 667 | 68 | |
XM_012957176.2 | XP_012812630.1 | 7 - 8 | 991 | 68 | ||
XM_012957177.2 | XP_012812631.1 | 6 - 7 | 846 | 68 | ||
XM_018091322.1 | XP_017946811.1 | 7 - 8 | 987 | 68 | ||
XM_018091323.1 | XP_017946812.1 | 6 - 7 | 690 | 68 | ||
GenBank mRNA | CR762036.2 | - | 5 - 6 | 667 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |