Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: brix1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045754.2 | NP_001039219.2 | 2 - 3 | 251 | 64 | |
GenBank mRNA | BC158497.1 | - | 2 - 3 | 251 | 64 | |
CR942614.2 | - | 2 - 3 | 271 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |