Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lancl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045803.1 | NP_001039268.1 | 5 - 6 | 810 | 77 | |
XM_012964858.2 | XP_012820312.1 | 6 - 7 | 1021 | 77 | ||
XM_012964859.2 | XP_012820313.1 | 7 - 8 | 1159 | 77 | ||
XM_018094689.1 | XP_017950178.1 | 7 - 8 | 1087 | 77 | ||
GenBank mRNA | BC135988.1 | - | 7 - 8 | 1137 | 77 | |
CR942790.2 | - | 5 - 6 | 810 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |