Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mettl7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078727.1 | NP_001072195.1 | 1 - 2 | 537 | 63 | |
GenBank mRNA | BC118737.1 | - | 1 - 2 | 537 | 63 | |
BC157212.1 | - | 1 - 2 | 504 | 63 | ||
CT027845.1 | - | 1 - 2 | 513 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |