Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cwc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078733.1 | NP_001072201.1 | 3 - 4 | 288 | 73 | |
XM_012957216.2 | XP_012812670.1 | 3 - 4 | 360 | 73 | ||
GenBank mRNA | BC118751.1 | - | 3 - 4 | 288 | 73 | |
BC158969.1 | - | 3 - 4 | 280 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |