Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nkain3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078767.1 | NP_001072235.1 | 2 - 3 | 441 | 69 | |
GenBank mRNA | BC118822.1 | - | 2 - 3 | 441 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |