Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frmd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018096974.1 | XP_017952463.1 | 1 - 2 | 313 | 103 | |
GenBank mRNA | BC118833.1 | - | 1 - 2 | 67 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |