Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078793.1 | NP_001072261.1 | 1 - 2 | 297 | 108 | |
GenBank mRNA | BC121249.1 | - | 1 - 2 | 297 | 108 | |
CT027965.3 | - | 1 - 2 | 295 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |