Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078814.1 | NP_001072282.1 | 3 - 4 | 643 | 97 | |
XM_018096301.1 | XP_017951790.1 | 3 - 4 | 504 | 97 | ||
GenBank mRNA | BC121300.1 | - | 3 - 4 | 643 | 97 | |
BC135462.1 | - | 3 - 4 | 583 | 97 | ||
CR762328.2 | - | 3 - 4 | 611 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |