Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078817.1 | NP_001072285.1 | 3 - 4 | 1387 | 69 | |
XM_012952548.2 | XP_012808002.1 | 3 - 4 | 1956 | 69 | ||
GenBank mRNA | BC121308.1 | - | 3 - 4 | 1387 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |