Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bicdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078860.1 | NP_001072328.1 | 7 - 8 | 1561 | 84 | |
GenBank mRNA | BC121401.1 | - | 7 - 8 | 1561 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |