Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdca9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078925.1 | NP_001072393.1 | 4 - 5 | 453 | 105 | |
GenBank mRNA | BC121590.1 | - | 4 - 5 | 453 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |