Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc10a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078948.1 | NP_001072416.1 | 1 | 1554 | 73 | |
GenBank mRNA | BC121646.1 | - | 1 | 1554 | 73 | |
CU075526.1 | - | 1 | 1364 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |