Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079148.1 | NP_001072616.1 | 10 - 11 | 1182 | 77 | |
XM_012961357.2 | XP_012816811.1 | 11 - 12 | 1322 | 77 | ||
XM_012961358.2 | XP_012816812.1 | 10 - 11 | 1259 | 77 | ||
GenBank mRNA | BC123006.1 | - | 10 - 11 | 1182 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |