Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079171.1 | NP_001072639.1 | 4 - 5 | 681 | 92 | |
GenBank mRNA | BC123912.1 | - | 4 - 5 | 681 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |