Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: art1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079214.1 | NP_001072682.1 | 112 | 110 | ||
GenBank mRNA | BC124013.1 | - | 112 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis |