Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nadk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079219.1 | NP_001072687.1 | 1 - 2 | 302 | 76 | |
XM_012965802.1 | XP_012821256.1 | 1 - 2 | 357 | 76 | ||
XM_012965806.2 | XP_012821260.1 | 1 - 2 | 360 | 76 | ||
XM_018094605.1 | XP_017950094.1 | 1 - 2 | 55 | 76 | ||
GenBank mRNA | BC124020.1 | - | 1 - 2 | 302 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |